Assay Details
| Gene Symbol: Cdkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183294.2 | NP_899117.1 | 6 - 7 | 834 | 109 | |
| GenBank mRNA | AK016781.1 | - | 6 - 7 | 835 | 109 | |
| BC115909.1 | - | 5 - 6 | 782 | 40105122 | 109 | |
| BC118055.1 | - | 5 - 6 | 783 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdkl1
Genomic Map