Assay Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021605.3 | NP_067618.1 | 3 - 4 | 357 | 108 | |
| GenBank mRNA | AF217650.1 | - | 2 - 3 | 208 | 108 | |
| AK035502.1 | - | 3 - 4 | 356 | 108 | ||
| AK078639.1 | - | 3 - 4 | 366 | 108 | ||
| AK088173.1 | - | 3 - 4 | 406 | 108 | ||
| AK149308.1 | - | 3 - 4 | 405 | 108 | ||
| AK165583.1 | - | 3 - 4 | 335 | 108 | ||
| BC026541.1 | - | 3 - 4 | 349 | 108 | ||
| BC037697.1 | - | 3 - 4 | 334 | 3994871 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek7
Genomic Map