Assay Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021605.3 | NP_067618.1 | 5 - 6 | 538 | 154 | |
| GenBank mRNA | AF217650.1 | - | 4 - 5 | 389 | 154 | |
| AK035502.1 | - | 5 - 6 | 537 | 154 | ||
| AK078639.1 | - | 5 - 6 | 547 | 154 | ||
| AK088173.1 | - | 5 - 6 | 587 | 154 | ||
| AK149308.1 | - | 5 - 6 | 586 | 154 | ||
| AK165583.1 | - | 5 - 6 | 516 | 154 | ||
| BC026541.1 | - | 5 - 6 | 530 | 154 | ||
| BC037697.1 | - | 5 - 6 | 515 | 3994871 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek7
Genomic Map