Assay Details
| Gene Symbol: Axdnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134071.1 | - | 8 - 9 | 959 | 74 | |
| BC150932.1 | - | 7 - 8 | 811 | 9087843 | 74 | |
| BC172088.1 | - | 7 - 8 | 901 | 9054374 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Axdnd1
Genomic Map