Assay Details
| Gene Symbol: Axdnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134071.1 | - | 2 - 3 | 202 | 89 | |
| BC150932.1 | - | 1 - 2 | 54 | 9087843 | 89 | |
| BC172088.1 | - | 1 - 2 | 54 | 9054374 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134071.1, BC150932.1, BC172088.1 transcripts.
Set Membership: |
|

Axdnd1
Genomic Map