Assay Details
| Gene Symbol: Axdnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134071.1 | - | 3 - 4 | 309 | 78 | |
| BC150932.1 | - | 2 - 3 | 161 | 9087843 | 78 | |
| BC172088.1 | - | 2 - 3 | 161 | 9054374 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Axdnd1
Genomic Map