Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100026967.mm.1 |
NM_001024945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026967.mm.1 |
NM_001024945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026967.mm.1 |
NM_023268.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026967.mm.1 |
NM_023268.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026967.mm.1 |
TC0100003309.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026964.mm.1 |
NM_001024945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026964.mm.1 |
NM_001024945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026964.mm.1 |
NM_023268.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026964.mm.1 |
NM_023268.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026964.mm.1 |
TC0100003309.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026972.mm.1 |
NM_001024945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026972.mm.1 |
NM_001024945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026972.mm.1 |
NM_023268.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026972.mm.1 |
NM_023268.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026972.mm.1 |
TC0100003309.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026968.mm.1 |
NM_001024945.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026968.mm.1 |
NM_001024945.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026968.mm.1 |
NM_023268.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026968.mm.1 |
NM_023268.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026968.mm.1 |
TC0100003309.mm.1 |
MTA TC |