Assay Details
| Gene Symbol: Rgsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243223.1 | NP_001230152.1 | 5 - 6 | 489 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgsl1| Gene Symbol: Rgsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243223.1 | NP_001230152.1 | 5 - 6 | 489 | 71 | |
Set Membership: |
|