| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100015309.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015309.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015309.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015310.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015310.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015310.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015302.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015302.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015302.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015306.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015306.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015306.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015308.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015308.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015308.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015301.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015301.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015301.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015304.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015304.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015304.mm.1 |
TC1100001643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100015307.mm.1 |
NM_134028.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015307.mm.1 |
NM_134028.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100015307.mm.1 |
TC1100001643.mm.1 |
MTA TC |