Assay Details
| Gene Symbol: Cntnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016782.2 | NP_058062.2 | 14 - 15 | 2359 | 63 | |
| GenBank mRNA | AF039833.1 | - | 14 - 15 | 2369 | 63 | |
| AK154468.1 | - | 3120 | 63 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cntnap1
Genomic Map