Assay Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146030.2 | NP_666142.1 | 10 - 11 | 1968 | 127 | |
| GenBank mRNA | AK052371.1 | - | 2 - 3 | 168 | 127 | |
| AK154510.1 | - | 10 - 11 | 1975 | 127 | ||
| AK164128.1 | - | 10 - 11 | 1978 | 127 | ||
| BC020025.1 | - | 10 - 11 | 1968 | 3490992 | 127 | |
| BC024538.1 | - | 3 - 4 | 396 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekhh3
Genomic Map