Assay Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146030.2 | NP_666142.1 | 2 - 3 | 640 | 60 | |
| GenBank mRNA | AK154510.1 | - | 2 - 3 | 656 | 60 | |
| AK164128.1 | - | 2 - 3 | 656 | 60 | ||
| BC020025.1 | - | 2 - 3 | 640 | 3490992 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhh3
Genomic Map