Assay Details
| Gene Symbol: Naa15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053089.3 | NP_444319.3 | 17 - 18 | 2466 | 79 | |
| GenBank mRNA | AB221683.1 | - | 17 - 18 | 2159 | 79 | |
| AF237622.1 | - | 13 - 14 | 1750 | 79 | ||
| AF510858.1 | - | 17 - 18 | 2356 | 79 | ||
| BC030167.1 | - | 5 - 6 | 674 | 79 | ||
| BC050017.1 | - | 17 - 18 | 2357 | 30020858 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa15
Genomic Map