Assay Details
| Gene Symbol: Naa15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053089.3 | NP_444319.3 | 3 - 4 | 547 | 121 | |
| GenBank mRNA | AB221683.1 | - | 3 - 4 | 240 | 121 | |
| AF510858.1 | - | 3 - 4 | 437 | 121 | ||
| BC050017.1 | - | 3 - 4 | 438 | 30020858 | 121 | |
| BQ887090.1 | - | 3 - 4 | 543 | 121 | ||
| CA327064.1 | - | 3 - 4 | 524 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naa15
Genomic Map