Assay Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010475.1 | NP_034605.1 | 2 - 3 | 294 | 77 | |
| GenBank mRNA | AK143346.1 | - | 2 - 3 | 243 | 77 | |
| BC125657.1 | - | 2 - 3 | 293 | 40130172 | 77 | |
| BC125659.1 | - | 2 - 3 | 293 | 40130174 | 77 | |
| X89627.1 | - | 2 - 3 | 294 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b1
Genomic Map