Assay Details
| Gene Symbol: Coasy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF521095.1 | - | 7 - 8 | 1415 | 62 | |
| AK004878.1 | - | 7 - 8 | 1475 | 62 | ||
| AK159405.1 | - | 7 - 8 | 1464 | 62 | ||
| AK166894.1 | - | 6 - 7 | 1758 | 62 | ||
| AK170330.1 | - | 6 - 7 | 1773 | 62 | ||
| BC020046.1 | - | 6 - 7 | 885 | 62 | ||
| BC064753.1 | - | 6 - 7 | 1403 | 62 | ||
More Information
Set Membership: |
|

Coasy
Genomic Map