Assay Details
| Gene Symbol: Sohlh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028937.3 | NP_083213.2 | 4 - 5 | 455 | 72 | |
| GenBank mRNA | AK016704.1 | - | 4 - 5 | 457 | 72 | |
| BC120917.1 | - | 4 - 5 | 450 | 40092728 | 72 | |
| BC120918.2 | - | 4 - 5 | 450 | 40092729 | 72 | |
| DQ086115.1 | - | 4 - 5 | 455 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sohlh2
Genomic Map