Assay Details
| Gene Symbol: Cpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007753.2 | NP_031779.1 | 1 - 2 | 76 | 87 | |
| GenBank mRNA | AK088906.1 | - | 1 - 2 | 77 | 87 | |
| BC108409.1 | - | 1 - 2 | 65 | 87 | ||
| J05118.1 | - | 1 - 2 | 72 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpa3
Genomic Map