Assay Details
| Gene Symbol: Cpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007753.2 | NP_031779.1 | 4 - 5 | 381 | 103 | |
| GenBank mRNA | AK088906.1 | - | 4 - 5 | 382 | 103 | |
| BC108409.1 | - | 4 - 5 | 370 | 103 | ||
| J05118.1 | - | 4 - 5 | 377 | 103 | ||
More Information
Set Membership: |
|

Cpa3
Genomic Map