Assay Details
| Gene Symbol: Cpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029706.1 | NP_083982.1 | 6 - 7 | 650 | 71 | |
| GenBank mRNA | AK003061.1 | - | 6 - 7 | 589 | 71 | |
| AK008678.1 | - | 3 - 4 | 322 | 71 | ||
| BC138759.1 | - | 6 - 7 | 608 | 8861781 | 71 | |
| BC138762.1 | - | 6 - 7 | 608 | 8861784 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpb1
Genomic Map