Assay Details
| Gene Symbol: Ctage5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165253.1 | NP_001158725.1 | 10 - 11 | 963 | 105 | |
| NM_001165254.1 | NP_001158726.1 | 10 - 11 | 962 | 105 | ||
| NM_146034.3 | NP_666146.3 | 12 - 13 | 1061 | 105 | ||
| GenBank mRNA | AK029466.1 | - | 10 - 11 | 962 | 105 | |
| AK053010.1 | - | 11 - 12 | 968 | 105 | ||
| AK087188.1 | - | 11 - 12 | 976 | 105 | ||
| AK157358.1 | - | 10 - 11 | 964 | 105 | ||
| BC018567.1 | - | 4 - 5 | 344 | 105 | ||
| BC024076.1 | - | 3 - 4 | 301 | 105 | ||
| BC026864.1 | - | 12 - 13 | 1019 | 2647214 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctage5
Genomic Map