Assay Details
| Gene Symbol: Ctage5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165253.1 | NP_001158725.1 | 8 - 9 | 813 | 76 | |
| NM_001165254.1 | NP_001158726.1 | 8 - 9 | 812 | 76 | ||
| NM_146034.3 | NP_666146.3 | 10 - 11 | 911 | 76 | ||
| GenBank mRNA | AK029466.1 | - | 8 - 9 | 812 | 76 | |
| AK053010.1 | - | 9 - 10 | 818 | 76 | ||
| AK087188.1 | - | 9 - 10 | 826 | 76 | ||
| AK157358.1 | - | 8 - 9 | 814 | 76 | ||
| BC018567.1 | - | 2 - 3 | 188 | 76 | ||
| BC024076.1 | - | 1 - 2 | 145 | 76 | ||
| BC026864.1 | - | 10 - 11 | 863 | 2647214 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctage5
Genomic Map