Assay Details
| Gene Symbol: Sec23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009147.2 | NP_033173.2 | 8 - 9 | 1177 | 86 | |
| GenBank mRNA | AY082671.1 | - | 8 - 9 | 1051 | 86 | |
| BC034610.1 | - | 8 - 9 | 1177 | 5364796 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec23a
Genomic Map