Assay Details
| Gene Symbol: Dld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007861.5 | NP_031887.2 | 7 - 8 | 711 | 114 | |
| GenBank mRNA | AK117104.1 | - | 7 - 8 | 655 | 114 | |
| AK136193.1 | - | 7 - 8 | 655 | 114 | ||
| AK153399.1 | - | 7 - 8 | 655 | 114 | ||
| AK167885.1 | - | 7 - 8 | 655 | 114 | ||
| AK168157.1 | - | 7 - 8 | 676 | 114 | ||
| AK168875.1 | - | 7 - 8 | 596 | 114 | ||
| AK193031.1 | - | 1 - 2 | 111 | 114 | ||
| U73445.1 | - | 7 - 8 | 688 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dld
Genomic Map