Assay Details
| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC048710.1 | - | 11 - 12 | 1412 | 6506827 | 138 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkar2b| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC048710.1 | - | 11 - 12 | 1412 | 6506827 | 138 |
Set Membership: |
|