Assay Details
| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011158.3 | NP_035288.2 | 2 - 3 | 536 | 89 | |
| GenBank mRNA | AK011647.1 | - | 2 - 3 | 494 | 89 | |
| AK041013.1 | - | 2 - 3 | 510 | 89 | ||
| AK135621.1 | - | 2 - 3 | 538 | 89 | ||
| AK138963.1 | - | 2 - 3 | 502 | 89 | ||
| BC048710.1 | - | 2 - 3 | 492 | 6506827 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkar2b
Genomic Map