Assay Details
| Gene Symbol: Prkar2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011158.3 | NP_035288.2 | 3 - 4 | 582 | 101 | |
| GenBank mRNA | AK011647.1 | - | 3 - 4 | 540 | 101 | |
| AK041013.1 | - | 3 - 4 | 556 | 101 | ||
| AK081805.1 | - | 3 - 4 | 359 | 101 | ||
| AK135621.1 | - | 3 - 4 | 584 | 101 | ||
| AK138963.1 | - | 3 - 4 | 548 | 101 | ||
| BC048710.1 | - | 3 - 4 | 538 | 6506827 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK081805.1 transcripts.
Set Membership: |
|

Prkar2b
Genomic Map