Assay Details
| Gene Symbol: Mccc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023644.4 | NP_076133.3 | 11 - 12 | 1372 | 68 | |
| GenBank mRNA | AF310338.1 | - | 11 - 12 | 1293 | 68 | |
| AK007782.1 | - | 11 - 12 | 1325 | 68 | ||
| AK031072.1 | - | 11 - 12 | 1333 | 68 | ||
| AK032203.1 | - | 11 - 12 | 1315 | 68 | ||
| AK160184.1 | - | 9 - 10 | 1089 | 68 | ||
| AK168589.1 | - | 11 - 12 | 1261 | 68 | ||
| BC021382.1 | - | 11 - 12 | 1336 | 5042993 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mccc1
Genomic Map