Assay Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173740.3 | NP_776101.3 | 3 - 4 | 444 | 98 | |
| GenBank mRNA | AK004337.1 | - | 3 - 4 | 306 | 98 | |
| AK146612.1 | - | 3 - 4 | 445 | 98 | ||
| AK155052.1 | - | 3 - 4 | 307 | 98 | ||
| AK164457.1 | - | 3 - 4 | 281 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Maoa
Genomic Map