Assay Details
| Gene Symbol: Tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011650.3 | NP_035780.1 | 4 - 5 | 438 | 74 | |
| GenBank mRNA | AF234179.1 | - | 4 - 5 | 438 | 74 | |
| AK010737.1 | - | 4 - 5 | 436 | 74 | ||
| AK150529.1 | - | 4 - 5 | 487 | 74 | ||
| AK165598.1 | - | 4 - 5 | 478 | 74 | ||
| AK201202.1 | - | 1 - 2 | 81 | 74 | ||
| AK219078.1 | - | 1 - 2 | 99 | 74 | ||
| BC004615.1 | - | 4 - 5 | 484 | 3585366 | 74 | |
| X81464.1 | - | 4 - 5 | 438 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tsn
Genomic Map