Assay Details
| Gene Symbol: Nifk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026472.4 | NP_080748.3 | 1 - 2 | 155 | 97 | |
| GenBank mRNA | AB056870.1 | - | 1 - 2 | 105 | 97 | |
| AK009440.1 | - | 1 - 2 | 122 | 97 | ||
| AK077770.1 | - | 1 - 2 | 150 | 97 | ||
| AK081486.1 | - | 1 - 2 | 122 | 97 | ||
| AY030275.1 | - | 1 - 2 | 126 | 97 | ||
| BC058559.1 | - | 1 - 2 | 134 | 6314031 | 97 | |
| BY289326.1 | - | 1 - 2 | 160 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nifk
Genomic Map