Assay Details
| Gene Symbol: Nifk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026472.4 | NP_080748.3 | 5 - 6 | 671 | 75 | |
| GenBank mRNA | AB056870.1 | - | 5 - 6 | 621 | 75 | |
| AK009440.1 | - | 5 - 6 | 638 | 75 | ||
| AK077770.1 | - | 5 - 6 | 666 | 75 | ||
| AK081486.1 | - | 5 - 6 | 638 | 75 | ||
| AY030275.1 | - | 5 - 6 | 642 | 75 | ||
| BC058559.1 | - | 5 - 6 | 650 | 6314031 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nifk
Genomic Map