Assay Details
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027941.1 | NP_082217.1 | 2 - 3 | 272 | 151 | |
| GenBank mRNA | AK005720.1 | - | 2 - 3 | 324 | 151 | |
| BC100519.1 | - | 2 - 3 | 272 | 30919449 | 151 | |
| BC116419.1 | - | 1 - 2 | 105 | 151 | ||
| BC116420.1 | - | 1 - 2 | 112 | 40103399 | 151 | |
| JF934738.1 | - | 2 - 3 | 223 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc34
Genomic Map