Assay Details
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027941.1 | NP_082217.1 | 8 - 9 | 922 | 99 | |
| GenBank mRNA | AK005720.1 | - | 8 - 9 | 974 | 99 | |
| BC100519.1 | - | 8 - 9 | 922 | 30919449 | 99 | |
| BC116419.1 | - | 7 - 8 | 755 | 99 | ||
| BC116420.1 | - | 7 - 8 | 762 | 40103399 | 99 | |
| JF934738.1 | - | 8 - 9 | 873 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc34
Genomic Map