Assay Details
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027941.1 | NP_082217.1 | 9 - 10 | 1079 | 67 | |
| GenBank mRNA | AK005720.1 | - | 9 - 10 | 1131 | 67 | |
| BC100519.1 | - | 9 - 10 | 1079 | 30919449 | 67 | |
| BC116419.1 | - | 8 - 9 | 912 | 67 | ||
| BC116420.1 | - | 8 - 9 | 919 | 40103399 | 67 | |
| JF934738.1 | - | 9 - 10 | 1030 | 67 | ||
More Information
Set Membership: |
|

Lrrc34
Genomic Map