Assay Details
| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008857.3 | NP_032883.2 | 1 - 2 | 314 | 121 | |
| GenBank mRNA | AK144979.1 | - | 1 - 2 | 314 | 121 | |
| AK147710.1 | - | 1 - 2 | 286 | 121 | ||
| AK167411.1 | - | 1 - 2 | 283 | 121 | ||
| AK220517.1 | - | 1 - 2 | 338 | 121 | ||
| BC021630.1 | - | 1 - 2 | 185 | 3980628 | 121 | |
| D28577.1 | - | 1 - 2 | 109 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144979.1, AK167411.1, BC021630.1, D28577.1 transcripts.
Set Membership: |
|

Prkci
Genomic Map