Assay Details
| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008857.3 | NP_032883.2 | 9 - 10 | 1092 | 124 | |
| GenBank mRNA | AK147710.1 | - | 9 - 10 | 1064 | 124 | |
| AK167411.1 | - | 9 - 10 | 1061 | 124 | ||
| AK220517.1 | - | 9 - 10 | 1116 | 124 | ||
| BC021630.1 | - | 9 - 10 | 963 | 3980628 | 124 | |
| D28577.1 | - | 9 - 10 | 887 | 124 | ||
More Information
Set Membership: |
|

Prkci
Genomic Map