Assay Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145800.1 | NP_001139272.1 | 12 - 13 | 1766 | 67 | |
| NM_033608.3 | NP_291086.2 | 12 - 13 | 1832 | 67 | ||
| GenBank mRNA | AF317839.1 | - | 12 - 13 | 1832 | 67 | |
| AK078536.1 | - | 12 - 13 | 1760 | 67 | ||
| AK145121.1 | - | 12 - 13 | 1741 | 67 | ||
| BC040281.1 | - | 12 - 13 | 1792 | 5364073 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf9
Genomic Map