Assay Details
| Gene Symbol: Cdhr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024478.1 | NP_001019649.1 | 6 - 7 | 817 | 100 | |
| GenBank mRNA | AK047666.1 | - | 6 - 7 | 818 | 100 | |
| AK049806.1 | - | 6 - 7 | 819 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdhr3
Genomic Map