Assay Details
| Gene Symbol: Tomm20l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029227.1 | NP_083503.1 | 2 - 3 | 196 | 118 | |
| GenBank mRNA | AK016101.1 | - | 2 - 3 | 181 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm20l
Genomic Map