Assay Details
| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134250.2 | NP_599011.2 | 1 - 2 | 126 | 72 | |
| GenBank mRNA | AF399831.1 | - | 1 - 2 | 72 | 72 | |
| AY553334.1 | - | 1 - 2 | 119 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Havcr2
Genomic Map