Assay Details
| Gene Symbol: Slc26a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177243.4 | NP_796217.2 | 18 - 19 | 2224 | 76 | |
| GenBank mRNA | AK078966.1 | - | 18 - 19 | 2225 | 76 | |
| AK086815.1 | - | 18 - 19 | 2225 | 76 | ||
| AY034145.1 | - | 17 - 18 | 2137 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a9
Genomic Map