Assay Details
| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008067.4 | NP_032093.3 | 3 - 4 | 540 | 81 | |
| GenBank mRNA | AK039144.1 | - | 3 - 4 | 481 | 81 | |
| AK134760.1 | - | 3 - 4 | 449 | 81 | ||
| BC119317.1 | - | 3 - 4 | 338 | 8734066 | 81 | |
| BC120686.1 | - | 3 - 4 | 338 | 40129609 | 81 | |
| M86568.1 | - | 3 - 4 | 442 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabra3
Genomic Map