Assay Details
| Gene Symbol: Olfr418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146651.2 | NP_666862.2 | 1 | 492 | 79 | |
| GenBank mRNA | BC119223.1 | - | 1 | 492 | 8733972 | 79 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142176 | BC119223.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140078 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142176 | BC119223.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P360092 | BC119223.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P360092 | NM_146651.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20836993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20837022 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140078 | - | - |
| Whole Mouse Genome G4122A | A_51_P360092 | BC119223.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360092 | NM_146651.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127858 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00823965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00823943 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00823949 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00823950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20837012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00085331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00706226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00783648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00783644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00823945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20837003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20836993 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351858 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351865 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351857 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360546 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2690584 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2690584 | - | - |
| mouse-6 v1.1 (Illumina) | scl14325.1.1_39-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360546 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14325.1.1_39-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2690584 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2690584 | - | - |

Olfr418
Genomic Map