Assay Details
| Gene Symbol: Actrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029690.2 | NP_083966.1 | 2 | 1763 | 151 | |
| GenBank mRNA | AK047260.1 | - | 2 | 1764 | 151 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029199 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029203 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029200 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029201 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029171 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039245 | NM_029690.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039245 | AK047260.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 465903 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177302 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177633 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039245 | NM_029690.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039245 | AK047260.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22029188 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177633 | - | - |
| Whole Mouse Genome G4122A | A_52_P126728 | BC107278.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P126728 | BC107277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P126728 | NM_029690.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P126728 | AK007208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P126728 | AK047260.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177633 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03082528 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02816683 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02598923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03082531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02598925 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02598922 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02816680 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02816682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02816681 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02598929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029200 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029201 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029171 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029202 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029203 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22029199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22029160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03082527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02757238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02975458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03103570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02514729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02463072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02816682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02816681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02991909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22029186 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10497620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497618 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429788_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429789_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10497617 | - | - |
| Mouse Genome 430 2.0 Array | 1429789_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429788_at | - | - |
| Mouse Genome 430 2.0 Array | 1429788_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429789_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76652_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76652_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300013804.mm.1 | NM_029690.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300013804.mm.1 | NM_029690.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300013804.mm.1 | TC0300001786.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300013805.mm.1 | NM_029690.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300013805.mm.1 | NM_029690.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300013805.mm.1 | TC0300001786.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300013806.mm.1 | NM_029690.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300013806.mm.1 | NM_029690.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300013806.mm.1 | TC0300001786.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429788_PM_at | - | - |
| Mouse Expression Set 430 | 1429789_at | - | - |
| Mouse Expression Set 430 | 1429789_PM_at | - | - |
| Mouse Expression Set 430 | 1429788_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429788_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429789_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429789_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429788_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 664039 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2570280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2729052 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2729052 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2729052 | - | - |

Actrt3
Genomic Map