Assay Details
| Gene Symbol: Ferd3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033522.2 | NP_277057.1 | 1 | 506 | 85 | |
| GenBank mRNA | AF517121.1 | - | 1 | 506 | 85 | |
| BC111576.1 | - | 1 | 498 | 40039965 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
floor plate development
negative regulation of transcription, DNA-templated
cell development
regulation of neurogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
protein dimerization activity
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
floor plate development
negative regulation of transcription, DNA-templated
cell development
regulation of neurogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977220 | NM_033522.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977220 | AF517121.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889036 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156409 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977220 | NM_033522.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977220 | AF517121.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28344127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156409 | - | - |
| Whole Mouse Genome G4122A | A_51_P276753 | NM_033522.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276753 | AF517121.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276753 | BC111576.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123180 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116137 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156409 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10132286 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10132292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10132283 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10132297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10132287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10132291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28344144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10099577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10293512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10099573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10374799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10374795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10435745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10371741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10102140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09842153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09943254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28344108 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395318 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422410_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395317 | - | - |
| Mouse Genome 430 2.0 Array | 1422410_at | - | - |
| Mouse Genome 430 2.0 Array | 1422410_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 114712_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 114712_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200002285.mm.1 | NM_033522.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200002285.mm.1 | NM_033522.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200002285.mm.1 | TC1200000296.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422410_at | - | - |
| Mouse Expression Set 430 | 1422410_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422410_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422410_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422410_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422410_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 856770 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3850673 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724181 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724181 | - | - |
| mouse-6 v1.1 (Illumina) | scl43292.1.1_224-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43292.1.1_224-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2724181 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2724181 | - | - |

Ferd3l
Genomic Map