Assay Details
| Gene Symbol: Nxpe5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC106195.1 | - | 132 | 6396133 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC106195.1 transcripts.
Set Membership: |
|

Nxpe5
Genomic Map