Assay Details
| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159743.1 | NP_001153215.1 | 1 - 2 | 713 | 107 | |
| GenBank mRNA | AK053903.1 | - | 1 - 2 | 597 | 107 | |
| AK134499.1 | - | 1 - 2 | 716 | 107 | ||
| BC046522.1 | - | 1 - 2 | 696 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam150b
Genomic Map