Assay Details
| Gene Symbol: Mybph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016749.2 | NP_058029.2 | 3 - 4 | 581 | 98 | |
| GenBank mRNA | BC116684.1 | - | 3 - 4 | 548 | 40106746 | 98 |
| BC118050.1 | - | 3 - 4 | 548 | 40106744 | 98 | |
| U68267.1 | - | 3 - 4 | 575 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mybph
Genomic Map