Assay Details
| Gene Symbol: Mybph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016749.2 | NP_058029.2 | 6 - 7 | 1008 | 59 | |
| GenBank mRNA | BC116684.1 | - | 6 - 7 | 975 | 40106746 | 59 |
| BC118050.1 | - | 6 - 7 | 975 | 40106744 | 59 | |
| CB587371.1 | - | 2 - 3 | 255 | 59 | ||
| U68267.1 | - | 6 - 7 | 1002 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mybph
Genomic Map